Assay Details
| Gene Symbol: Irf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013674.1 | NP_038702.1 | 4 - 5 | 564 | 77 | |
| GenBank mRNA | AK089819.1 | - | 4 - 5 | 586 | 77 | |
| AK154138.1 | - | 4 - 5 | 585 | 77 | ||
| AK154315.1 | - | 4 - 5 | 576 | 77 | ||
| AK156562.1 | - | 4 - 5 | 586 | 77 | ||
| BC137713.1 | - | 4 - 5 | 552 | 8860732 | 77 | |
| BC137714.1 | - | 4 - 5 | 552 | 8860733 | 77 | |
| U11692.1 | - | 4 - 5 | 564 | 77 | ||
| U34307.1 | - | 4 - 5 | 558 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Irf4
Genomic Map