Assay Details
| Gene Symbol: Trpv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022413.4 | NP_071858.3 | 11 - 12 | 1738 | 64 | |
| GenBank mRNA | AJ542487.1 | - | 11 - 12 | 1443 | 64 | |
| AK077234.1 | - | 11 - 12 | 1668 | 64 | ||
| AK168760.1 | - | 11 - 12 | 1738 | 64 | ||
| BC016101.1 | - | 11 - 12 | 1676 | 4911355 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trpv6
Genomic Map