Assay Details
| Gene Symbol: Trpv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022413.4 | NP_071858.3 | 6 - 7 | 1058 | 94 | |
| GenBank mRNA | AB037373.1 | - | 6 - 7 | 962 | 94 | |
| AJ542487.1 | - | 6 - 7 | 763 | 94 | ||
| AK077234.1 | - | 6 - 7 | 988 | 94 | ||
| AK085123.1 | - | 4 - 5 | 1671 | 95 | ||
| AK168760.1 | - | 6 - 7 | 1058 | 94 | ||
| BC016101.1 | - | 6 - 7 | 996 | 4911355 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trpv6
Genomic Map