Assay Details
| Gene Symbol: Ephb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146351.1 | NP_001139823.1 | 11 - 12 | 2405 | 81 | |
| NM_007680.4 | NP_031706.3 | 11 - 12 | 2401 | 81 | ||
| GenBank mRNA | AK076442.1 | - | 11 - 12 | 2228 | 81 | |
| AK087423.1 | - | 11 - 12 | 2316 | 81 | ||
| AK163823.1 | - | 2 - 3 | 123 | 81 | ||
| BC031924.1 | - | 10 - 11 | 2013 | 4911959 | 81 | |
| L77867.1 | - | 11 - 12 | 2095 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephb6
Genomic Map