Assay Details
| Gene Symbol: Atf6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017406.4 | NP_059102.2 | 16 - 17 | 1884 | 79 | |
| GenBank mRNA | AK143765.1 | - | 16 - 17 | 1817 | 79 | |
| AK146671.1 | - | 16 - 17 | 1851 | 79 | ||
| AK159169.1 | - | 16 - 17 | 1808 | 79 | ||
| BC013534.1 | - | 16 - 17 | 1884 | 4239106 | 79 | |
| BC052635.1 | - | 16 - 17 | 1819 | 6395626 | 79 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Atf6b
Genomic Map