Assay Details
Gene Symbol: Olfr434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146369.1 | NP_666481.1 | 1 | 324 | 75 | |
GenBank mRNA | BC120611.1 | - | 1 | 356 | 40129534 | 75 |
BC120613.1 | - | 1 | 356 | 40129536 | 75 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
