Assay Details
| Gene Symbol: Hist1h2ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC147161.1 | - | 1 | 119 | 9056189 | 90 |
| BC147162.1 | - | 1 | 119 | 9056203 | 90 | |
| Gene Symbol: Hist1h2ao | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177544.2 | NP_001171015.1 | 1 | 119 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC147161.1, BC147162.1, NM_001177544.2 transcripts.
Set Membership: |
|

Hist1h2ap
Genomic Map