Assay Details
| Gene Symbol: Hist1h2ac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033518.1 | - | 548 | 136 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28882830 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28882829 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084691 | NM_178189.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084691 | AK033518.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215428 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160493 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166007 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171705 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084691 | NM_178189.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084691 | AK033518.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213597 | AK033518.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882829 | - | - |
| Whole Mouse Genome G4122A | A_51_P213597 | AK033518.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10491539 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10491537 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10491550 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10491558 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28882829 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28882830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11090366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11090365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11094400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10654294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10758204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10648747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10491555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10491552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28882829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10758203 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10408221 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408222 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408111 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408220 | - | - |
| Mouse Expression Set 430 | 1418366_PM_at | - | - |
| Mouse Expression Set 430 | 1438009_PM_at | - | - |
| Mouse Expression Set 430 | 1418366_at | - | - |
| Mouse Expression Set 430 | 1438009_at | - | - |
| Mouse Expression Set 430 | 1418367_x_at | - | - |
| Mouse Expression Set 430 | 1418367_PM_x_at | - | - |
| Murine Genome U74v2 Set | 94805_f_at | - | - |
| Murine Genome U74v2 Set | 94805_PM_f_at | - | - |
| Murine Genome U74v2 Set | 167053_at | - | - |
| Murine Genome U74v2 Set | 167053_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418367_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418366_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418366_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418367_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438009_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438009_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3130292 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3130292 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105290441 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003130292 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2666418 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2666418 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226839 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221342 | - | - |
| mouse-6 v1.1 (Illumina) | GI_31542957-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00319164.1_303-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3130292 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105290441 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00319164.1_303-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3130292 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105290441 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2666418 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2666418 | - | - |

Hist1h2ac
Genomic Map