Assay Details
| Gene Symbol: Lrguk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028886.1 | NP_083162.1 | 10 - 11 | 1225 | 80 | |
| GenBank mRNA | AK014976.1 | - | 10 - 11 | 1229 | 80 | |
| AK029666.1 | - | 10 - 11 | 1224 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrguk
Genomic Map