Assay Details
| Gene Symbol: Lrguk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028886.1 | NP_083162.1 | 17 - 18 | 2071 | 65 | |
| GenBank mRNA | AK014976.1 | - | 17 - 18 | 2075 | 65 | |
| AK029666.1 | - | 17 - 18 | 2069 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrguk
Genomic Map