Assay Details
| Gene Symbol: Lrguk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028886.1 | NP_083162.1 | 7 - 8 | 954 | 121 | |
| GenBank mRNA | AK014976.1 | - | 7 - 8 | 958 | 121 | |
| AK029666.1 | - | 7 - 8 | 953 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrguk
Genomic Map