Assay Details
| Gene Symbol: Tomm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025394.3 | NP_079670.1 | 2 - 3 | 228 | 79 | |
| GenBank mRNA | AK003864.1 | - | 2 - 3 | 191 | 79 | |
| AK008581.1 | - | 2 - 3 | 145 | 79 | ||
| BC055352.1 | - | 2 - 3 | 206 | 5011269 | 79 | |
| BU555283.1 | - | 2 - 3 | 231 | 79 | ||
| BY773505.1 | - | 2 - 3 | 221 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tomm7
Genomic Map