| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600002376.mm.1 |
TC0600000324.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600002376.mm.1 |
NM_008012.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002376.mm.1 |
NM_008012.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002374.mm.1 |
TC0600000324.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600002374.mm.1 |
NM_008012.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002374.mm.1 |
NM_008012.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002372.mm.1 |
TC0600000324.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600002372.mm.1 |
NM_008012.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002372.mm.1 |
NM_008012.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002375.mm.1 |
TC0600000324.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600002375.mm.1 |
NM_008012.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002375.mm.1 |
NM_008012.1 |
Refseq |