Assay Details
| Gene Symbol: Pon2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183308.2 | NP_899131.1 | 5 - 6 | 584 | 129 | |
| GenBank mRNA | AK007350.1 | - | 5 - 6 | 553 | 130 | |
| AK146311.1 | - | 5 - 6 | 552 | 129 | ||
| AK152351.1 | - | 5 - 6 | 535 | 129 | ||
| AK171926.1 | - | 5 - 6 | 542 | 129 | ||
| BC037140.1 | - | 5 - 6 | 526 | 5027719 | 129 | |
| BC055896.1 | - | 5 - 6 | 584 | 3482584 | 129 | |
| BC062200.1 | - | 5 - 6 | 575 | 6399484 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pon2
Genomic Map