Assay Details
Gene Symbol: Olfr479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011742.1 | NP_001011742.1 | 1 | 547 | 114 | |
GenBank mRNA | BC150837.1 | - | 1 | 574 | 9087748 | 114 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115960 | NM_001011742.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115960 | BC150837.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115960 | NM_001011742.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115960 | BC150837.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402105 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402099 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402117 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402118 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402084 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402082 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402124 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402103 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402080 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402088 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402094 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402122 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402096 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402086 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402106 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402075 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402107 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402121 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402125 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402101 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402113 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402085 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402098 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402091 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402127 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402072 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402100 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402093 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402069 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402120 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402089 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402109 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402092 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402134 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402094 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402122 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402117 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402118 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402093 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402103 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402086 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402099 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402079 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402129 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402082 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402101 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402091 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402127 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402131 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402106 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402096 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402098 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402100 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402120 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402136 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402109 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402134 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402089 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402088 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402125 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402130 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402084 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402128 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402124 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402113 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10556128 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10556127 | - | - |
Affymetrix Mouse Clariom S Array | PSR0700013266.mm.1 | TC0700001590.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700013268.mm.1 | TC0700001590.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700013267.mm.1 | TC0700001590.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700013267.mm.1 | NM_001011742.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700013267.mm.1 | NM_001011742.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 698943 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3162046 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_3162046 | - | - |
mouse-6 v1.1 (Illumina) | GI_58801277-S | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3162046 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3162046 | - | - |
