Assay Details
| Gene Symbol: Tac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009311.2 | NP_033337.1 | 4 - 5 | 367 | 118 | |
| GenBank mRNA | BC117081.1 | - | 4 - 5 | 342 | 40126400 | 118 |
| BC119122.1 | - | 4 - 5 | 342 | 8733871 | 118 | |
| D17584.1 | - | 4 - 5 | 367 | 118 | ||
| M68908.1 | - | 3 - 4 | 163 | 118 | ||
More Information
Set Membership: |
|

Tac1
Genomic Map