Assay Details
| Gene Symbol: Gdpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177696.3 | NP_808364.2 | 11 - 12 | 1186 | 92 | |
| GenBank mRNA | AK029527.1 | - | 11 - 12 | 1190 | 92 | |
| AK161500.1 | - | 11 - 12 | 1153 | 92 | ||
| BC140990.1 | - | 9 - 10 | 914 | 9055602 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gdpd4
Genomic Map