Assay Details
| Gene Symbol: Gdpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177696.3 | NP_808364.2 | 7 - 8 | 728 | 77 | |
| GenBank mRNA | AK029527.1 | - | 7 - 8 | 732 | 77 | |
| AK161500.1 | - | 7 - 8 | 695 | 77 | ||
| BC140990.1 | - | 5 - 6 | 456 | 9055602 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gdpd4
Genomic Map