Assay Details
| Gene Symbol: Olfr659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147050.1 | NP_667261.1 | 1 | 442 | 103 | |
| GenBank mRNA | BC148221.1 | - | 1 | 446 | 40136035 | 103 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr659
Genomic Map