Assay Details
| Gene Symbol: Slc26a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289787.1 | NP_001276716.1 | 10 - 11 | 1361 | 66 | |
| NM_001289788.1 | NP_001276717.1 | 10 - 11 | 1309 | 66 | ||
| NM_030727.5 | NP_109652.3 | 12 - 13 | 1547 | 66 | ||
| GenBank mRNA | AF529192.1 | - | 12 - 13 | 1547 | 66 | |
| AY024359.2 | - | 10 - 11 | 1313 | 66 | ||
| BC108986.1 | - | 10 - 11 | 1301 | 40047724 | 66 | |
| BC108987.1 | - | 10 - 11 | 1353 | 40047725 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a5
Genomic Map