Assay Details
| Gene Symbol: Dnajc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009584.4 | NP_033610.1 | 7 - 8 | 834 | 137 | |
| GenBank mRNA | AK131964.1 | - | 7 - 8 | 855 | 137 | |
| AK136951.1 | - | 7 - 8 | 858 | 137 | ||
| AK161347.1 | - | 7 - 8 | 801 | 137 | ||
| AK161376.1 | - | 7 - 8 | 855 | 137 | ||
| AK167991.1 | - | 7 - 8 | 855 | 137 | ||
| AK169850.1 | - | 7 - 8 | 869 | 137 | ||
| BC021755.1 | - | 7 - 8 | 839 | 137 | ||
| BC052027.1 | - | 7 - 8 | 861 | 5718637 | 137 | |
| D63784.1 | - | 7 - 8 | 833 | 137 | ||
| U53208.1 | - | 7 - 8 | 789 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc2
Genomic Map