Assay Details
| Gene Symbol: Mrvi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010826.5 | NP_034956.2 | 18 - 19 | 2481 | 60 | |
| NM_194464.3 | NP_919446.2 | 19 - 20 | 2724 | 60 | ||
| GenBank mRNA | AK141821.1 | - | 14 - 15 | 1763 | 60 | |
| U63407.1 | - | 19 - 20 | 2727 | 60 | ||
| U63408.1 | - | 18 - 19 | 2476 | 60 | ||
More Information
Set Membership: |
|

Mrvi1
Genomic Map