Assay Details
Gene Symbol: Tas2r137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025385.1 | NP_001020556.1 | 1 | 942 | 131 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
detection of chemical stimulus involved in sensory perception of bitter taste
signal transduction
G-protein coupled receptor signaling pathway
response to stimulus
sensory perception of taste
signal transducer activity
G-protein coupled receptor activity
taste receptor activity
bitter taste receptor activity
detection of chemical stimulus involved in sensory perception of bitter taste
signal transduction
G-protein coupled receptor signaling pathway
response to stimulus
sensory perception of taste
signal transducer activity
G-protein coupled receptor activity
taste receptor activity
bitter taste receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076041 | NM_001025385.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 820637 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076041 | NM_001025385.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389184 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389181 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389167 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389163 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389179 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389186 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389160 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389178 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389169 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389173 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389165 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389188 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389175 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389171 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389162 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389180 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389177 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24389158 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05413419 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05413424 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05413396 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05413407 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05283944 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05283941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389209 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24389139 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05358793 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389178 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389163 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389155 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389177 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389181 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389162 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389180 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389188 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389153 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389175 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389160 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389173 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05399228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389165 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389171 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389149 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389186 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389169 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389147 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05450179 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05280791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389183 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389150 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389152 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389189 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24389158 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10537500 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10537499 | - | - |
Affymetrix Mouse Clariom S Array | PSR0600003201.mm.1 | TC0600000412.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600003201.mm.1 | NM_001025385.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600003201.mm.1 | NM_001025385.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 826695 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160550 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_3160550 | - | - |
mouse-6 v1.1 (Illumina) | GI_71480159-S | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3160550 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160550 | - | - |
