Assay Details
| Gene Symbol: Sema3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013657.5 | NP_038685.3 | 2 - 3 | 293 | 79 | |
| GenBank mRNA | AK140348.1 | - | 829 | 79 | ||
| BC066852.1 | - | 2 - 3 | 293 | 30093230 | 79 | |
| X85994.1 | - | 2 - 3 | 245 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema3c
Genomic Map