Assay Details
| Gene Symbol: Peg10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040611.1 | NP_001035701.1 | 1 - 2 | 275 | 61 | |
| NM_130877.2 | NP_570947.2 | 1 - 2 | 275 | 61 | ||
| GenBank mRNA | AB091827.1 | - | 1 - 2 | 201 | 61 | |
| AJ006464.1 | - | 1 - 2 | 190 | 61 | ||
| CJ127881.1 | - | 1 - 2 | 275 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB091827.1, NM_001040611.1, NM_130877.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206541 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206537 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206535 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206534 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206542 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206536 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206540 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206533 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2029763 | NM_001040611.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2029763 | NM_130877.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2029763 | AB091827.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2029763 | AJ006464.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377723 | BC094558.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377723 | NM_001040611.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377723 | NM_130877.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377723 | AB091827.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377723 | BC094558.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377723 | NM_001040611.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377723 | NM_130877.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377723 | AB091827.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801153 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741186 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576970 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348236 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762031 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 796992 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 258457 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 138708 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377723 | BC094558.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377723 | NM_001040611.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377723 | NM_130877.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377723 | AB091827.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377723 | BC094558.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377723 | NM_001040611.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377723 | NM_130877.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377723 | AB091827.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2029763 | NM_001040611.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2029763 | NM_130877.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2029763 | AB091827.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2029763 | AJ006464.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206561 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24206569 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05797715 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01255919 | - | - |
| Whole Mouse Genome G4122A | A_52_P377723 | BC094558.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377723 | NM_001040611.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377723 | NM_130877.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377723 | AB091827.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377723 | BC094558.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377723 | NM_001040611.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377723 | NM_130877.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377723 | AB091827.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134852 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05797715 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05124711 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05124708 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206541 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206537 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206533 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206534 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206536 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206542 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206535 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24206540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24206626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05124705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206564 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05124709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05124706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05124700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05124708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05568782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206561 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05124713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206592 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05857714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05310490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05310489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05124711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05314930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24206580 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10536296 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536295 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427550_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10536294 | - | - |
| Mouse Genome 430 2.0 Array | 1427550_at | - | - |
| Mouse Genome 430 2.0 Array | 1427550_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170676_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170676_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600000190.mm.1 | TC0600000014.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600000190.mm.1 | NM_001040611.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000190.mm.1 | NM_001040611.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000190.mm.1 | NM_130877.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000190.mm.1 | NM_130877.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000188.mm.1 | TC0600000014.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600000188.mm.1 | NM_001040611.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000188.mm.1 | NM_001040611.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000188.mm.1 | NM_130877.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000188.mm.1 | NM_130877.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000191.mm.1 | TC0600000014.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600000191.mm.1 | NM_001040611.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000191.mm.1 | NM_001040611.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000191.mm.1 | NM_130877.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000191.mm.1 | NM_130877.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000189.mm.1 | TC0600000014.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600000189.mm.1 | NM_001040611.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000189.mm.1 | NM_001040611.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000189.mm.1 | NM_130877.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000189.mm.1 | NM_130877.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000194.mm.1 | TC0600000014.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600000194.mm.1 | NM_001040611.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000194.mm.1 | NM_001040611.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000194.mm.1 | NM_130877.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600000194.mm.1 | NM_130877.2 | Refseq |
| Mouse Expression Set 430 | 1427550_at | - | - |
| Mouse Expression Set 430 | 1427550_PM_at | - | - |
| Murine Genome U74v2 Set | 92269_PM_r_at | - | - |
| Murine Genome U74v2 Set | 92269_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427550_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427550_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427550_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427550_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 445222 | - | - |
| ABI Mouse Genome Survey Microarray | 751405 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100540538 | - | - |
| MouseWG-6 v2 Expression BeadChip | 104210161 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2517953 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3037645 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3037645 | - | - |

Peg10
Genomic Map