| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700001627.mm.1 |
NM_025934.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001627.mm.1 |
NM_025934.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001627.mm.1 |
TC1700000202.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001628.mm.1 |
NM_025934.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001628.mm.1 |
NM_025934.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001628.mm.1 |
TC1700000202.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001629.mm.1 |
NM_025934.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001629.mm.1 |
NM_025934.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001629.mm.1 |
TC1700000202.mm.1 |
MTA TC |