Assay Details
| Gene Symbol: Riok2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025934.2 | NP_080210.1 | 3 - 4 | 591 | 101 | |
| GenBank mRNA | AK008385.1 | - | 3 - 4 | 350 | 101 | |
| AK008451.1 | - | 3 - 4 | 351 | 101 | ||
| AK049232.1 | - | 3 - 4 | 335 | 101 | ||
| AK090011.1 | - | 3 - 4 | 354 | 101 | ||
| BC010781.1 | - | 3 - 4 | 332 | 4216633 | 101 | |
| BC080771.1 | - | 3 - 4 | 614 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Riok2
Genomic Map