Assay Details
| Gene Symbol: Lnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172827.3 | NP_766415.1 | 16 - 17 | 2942 | 137 | |
| GenBank mRNA | AK029094.1 | - | 16 - 17 | 2943 | 137 | |
| AK161484.1 | - | 2169 | 137 | |||
| BC120925.1 | - | 16 - 17 | 2899 | 40098425 | 137 | |
| BC120926.1 | - | 16 - 17 | 2899 | 40098428 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lnpep
Genomic Map