Assay Details
Gene Symbol: Olfr791 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146930.1 | NP_667141.1 | 1 | 843 | 110 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310741 | NM_146930.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310741 | NM_146930.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162479 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310741 | NM_146930.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310741 | NM_146930.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310741 | NM_146930.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310741 | NM_146930.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546185 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546188 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546181 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546180 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546170 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546195 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546186 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546160 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546177 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546182 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546179 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546193 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546161 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546164 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546184 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546155 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546168 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546163 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546173 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546165 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546156 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546172 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546176 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546174 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546196 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546157 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546159 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162479 | - | - |
Whole Mouse Genome G4122A | A_51_P310741 | NM_146930.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P310741 | NM_146930.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114131 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162479 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08894604 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08894617 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08894610 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08894613 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08894615 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08894621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546179 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546185 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08472139 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546161 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546164 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546177 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546196 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546159 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546155 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08894609 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546199 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546174 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546181 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08894612 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08894624 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546171 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546160 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546189 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546165 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08894610 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546176 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546170 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546163 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08780380 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546186 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546156 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27546157 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10367506 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10367505 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 848413 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 130095 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000130095 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2748377 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2748377 | - | - |
mouse-6 v1.1 (Illumina) | scl5600.1.1_237-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 130095 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl5600.1.1_237-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 130095 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2878106 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2878106 | - | - |
