Assay Details
| Gene Symbol: Dcstamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289506.1 | NP_001276435.1 | 1 - 2 | 38 | 61 | |
| NM_001289508.1 | NP_001276437.1 | 1 - 2 | 38 | 61 | ||
| NM_001289512.1 | NP_001276441.1 | 1 - 2 | 38 | 61 | ||
| NM_001289513.1 | NP_001276442.1 | 1 - 2 | 38 | 61 | ||
| NM_029422.4 | NP_083698.1 | 1 - 2 | 38 | 61 | ||
| GenBank mRNA | AB109560.1 | - | 1 - 2 | 36 | 61 | |
| AB109561.1 | - | 1 - 2 | 36 | 61 | ||
| AK014697.1 | - | 1 - 2 | 37 | 61 | ||
| AK154384.1 | - | 1 - 2 | 37 | 61 | ||
| AY517483.1 | - | 1 - 2 | 37 | 61 | ||
| AY517484.1 | - | 1 - 2 | 37 | 61 | ||
| AY517485.1 | - | 1 - 2 | 37 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB109560.1, AB109561.1, AK014697.1, AK154384.1, AY517483.1, AY517484.1, AY517485.1, NM_001289506.1, NM_001289508.1, NM_001289512.1, NM_001289513.1, NM_029422.4 transcripts.
Set Membership: |
|

Dcstamp
Genomic Map