Assay Details
| Gene Symbol: Dcstamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289512.1 | NP_001276441.1 | 2 - 3 | 1075 | 89 | |
| GenBank mRNA | AK014697.1 | - | 2 - 3 | 1074 | 89 | |
| AY517485.1 | - | 2 - 3 | 1074 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcstamp
Genomic Map