Assay Details
| Gene Symbol: Zc3hc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ537495.1 | - | 9 - 10 | 1440 | 67 | |
| AK151694.1 | - | 9 - 10 | 1444 | 67 | ||
| AK167465.1 | - | 9 - 10 | 1449 | 67 | ||
| AK168071.1 | - | 9 - 10 | 1458 | 67 | ||
| BC024560.1 | - | 3 - 4 | 464 | 67 | ||
| BC037445.1 | - | 9 - 10 | 1402 | 67 | ||
| BC050141.1 | - | 9 - 10 | 1441 | 30011212 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zc3hc1
Genomic Map