Assay Details
| Gene Symbol: Cpa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144537.3 | NP_653120.1 | 1 - 2 | 433 | 78 | |
| GenBank mRNA | AF466283.1 | - | 1 - 2 | 434 | 78 | |
| AK015256.1 | - | 1 - 2 | 434 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF466283.1, NM_144537.3 transcripts.
Set Membership: |
|

Cpa5
Genomic Map