| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600001714.mm.1 |
TC0600000245.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600001714.mm.1 |
NM_001171000.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001714.mm.1 |
NM_001171000.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001714.mm.1 |
NM_001171001.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001714.mm.1 |
NM_001171001.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001714.mm.1 |
NM_021414.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001714.mm.1 |
NM_021414.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001715.mm.1 |
TC0600000245.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600001715.mm.1 |
NM_001171000.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001715.mm.1 |
NM_001171000.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001715.mm.1 |
NM_001171001.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001715.mm.1 |
NM_001171001.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001715.mm.1 |
NM_021414.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001715.mm.1 |
NM_021414.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001717.mm.1 |
TC0600000245.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600001717.mm.1 |
NM_001171000.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001717.mm.1 |
NM_001171000.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001717.mm.1 |
NM_001171001.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001717.mm.1 |
NM_001171001.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001717.mm.1 |
NM_021414.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001717.mm.1 |
NM_021414.6 |
Refseq |