Assay Details
| Gene Symbol: Cpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025350.3 | NP_079626.2 | 2 - 3 | 381 | 70 | |
| GenBank mRNA | AK003088.1 | - | 2 - 3 | 166 | 70 | |
| BC052661.1 | - | 2 - 3 | 395 | 30050094 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpa1
Genomic Map