Assay Details
| Gene Symbol: Cpa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024698.2 | NP_001019869.1 | 1 - 2 | 93 | 88 | |
| GenBank mRNA | BC094929.1 | - | 1 - 2 | 123 | 6434004 | 88 |
More Information
Updated Mapping Information
Set Membership: |
|

Cpa2
Genomic Map