Assay Details
| Gene Symbol: Galntl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026449.3 | NP_080725.2 | 3 - 4 | 508 | 67 | |
| GenBank mRNA | AK005605.1 | - | 3 - 4 | 529 | 67 | |
| AK006193.1 | - | 3 - 4 | 487 | 67 | ||
| AK016415.1 | - | 3 - 4 | 509 | 67 | ||
| BC049554.1 | - | 3 - 4 | 490 | 67 | ||
| BC100447.1 | - | 3 - 4 | 361 | 67 | ||
| BC145757.1 | - | 3 - 4 | 368 | 40131060 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galntl5
Genomic Map