Assay Details
| Gene Symbol: Ubn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177185.4 | NP_796159.3 | 11 - 12 | 2179 | 72 | |
| GenBank mRNA | AK051609.1 | - | 11 - 12 | 1552 | 72 | |
| BC043095.1 | - | 1424 | 72 | |||
| BC044744.1 | - | 6 - 7 | 881 | 72 | ||
| BC051458.1 | - | 11 - 12 | 2154 | 6468017 | 72 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ubn2
Genomic Map