Assay Details
| Gene Symbol: Ubn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177185.4 | NP_796159.3 | 7 - 8 | 1668 | 115 | |
| GenBank mRNA | BC043095.1 | - | 913 | 115 | ||
| BC044744.1 | - | 2 - 3 | 370 | 115 | ||
| BC051458.1 | - | 7 - 8 | 1643 | 6468017 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubn2
Genomic Map