Assay Details
| Gene Symbol: Kdm7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033430.4 | NP_001028602.2 | 10 - 11 | 1479 | 104 | |
| GenBank mRNA | AK042327.1 | - | 10 - 11 | 1496 | 104 | |
| AK136209.1 | - | 10 - 11 | 1318 | 104 | ||
| AK136238.1 | - | 10 - 11 | 1466 | 104 | ||
| BC007161.1 | - | 10 - 11 | 1175 | 104 | ||
| BC145216.1 | - | 10 - 11 | 1365 | 104 | ||
| BC145848.1 | - | 10 - 11 | 1369 | 40131151 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdm7a
Genomic Map