Assay Details
| Gene Symbol: Kdm7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033430.4 | NP_001028602.2 | 2 - 3 | 421 | 60 | |
| GenBank mRNA | AK042327.1 | - | 2 - 3 | 438 | 60 | |
| AK042834.1 | - | 2 - 3 | 422 | 60 | ||
| AK136209.1 | - | 2 - 3 | 260 | 60 | ||
| AK136238.1 | - | 2 - 3 | 408 | 60 | ||
| BC007161.1 | - | 2 - 3 | 117 | 60 | ||
| BC145216.1 | - | 2 - 3 | 311 | 60 | ||
| BC145848.1 | - | 2 - 3 | 311 | 40131151 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdm7a
Genomic Map