Assay Details
| Gene Symbol: Kdm7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033430.4 | NP_001028602.2 | 9 - 10 | 1377 | 85 | |
| GenBank mRNA | AK042327.1 | - | 9 - 10 | 1394 | 85 | |
| AK042834.1 | - | 9 - 10 | 1378 | 85 | ||
| AK136209.1 | - | 9 - 10 | 1216 | 85 | ||
| AK136238.1 | - | 9 - 10 | 1364 | 85 | ||
| BC007161.1 | - | 9 - 10 | 1073 | 85 | ||
| BC145216.1 | - | 9 - 10 | 1263 | 85 | ||
| BC145848.1 | - | 9 - 10 | 1267 | 40131151 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdm7a
Genomic Map