Assay Details
| Gene Symbol: Parp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172893.3 | NP_766481.2 | 4 - 5 | 1089 | 102 | |
| GenBank mRNA | AK036886.1 | - | 4 - 5 | 1092 | 102 | |
| AK053205.1 | - | 4 - 5 | 990 | 102 | ||
| AK081528.1 | - | 4 - 5 | 990 | 102 | ||
| AK156623.1 | - | 4 - 5 | 958 | 102 | ||
| AK162200.1 | - | 4 - 5 | 660 | 102 | ||
| BC120733.1 | - | 4 - 5 | 922 | 40129656 | 102 | |
| BC137645.1 | - | 4 - 5 | 922 | 8860664 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parp12
Genomic Map