Assay Details
| Gene Symbol: Parp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172893.3 | NP_766481.2 | 5 - 6 | 1200 | 87 | |
| GenBank mRNA | AK036886.1 | - | 5 - 6 | 1203 | 87 | |
| AK053205.1 | - | 5 - 6 | 1101 | 87 | ||
| AK081528.1 | - | 5 - 6 | 1101 | 87 | ||
| AK156623.1 | - | 5 - 6 | 1069 | 87 | ||
| AK162200.1 | - | 5 - 6 | 771 | 87 | ||
| AK204869.1 | - | 2 - 3 | 137 | 87 | ||
| BC120733.1 | - | 5 - 6 | 1033 | 40129656 | 87 | |
| BC137645.1 | - | 5 - 6 | 1033 | 8860664 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parp12
Genomic Map