Assay Details
| Gene Symbol: Parp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172893.3 | NP_766481.2 | 8 - 9 | 1641 | 59 | |
| GenBank mRNA | AK036886.1 | - | 8 - 9 | 1644 | 59 | |
| AK053205.1 | - | 8 - 9 | 1542 | 59 | ||
| AK081528.1 | - | 8 - 9 | 1543 | 59 | ||
| AK156623.1 | - | 8 - 9 | 1510 | 59 | ||
| AK162200.1 | - | 8 - 9 | 1212 | 59 | ||
| AK218645.1 | - | 2 - 3 | 201 | 59 | ||
| BC024579.1 | - | 3 - 4 | 290 | 59 | ||
| BC048927.1 | - | 4 - 5 | 492 | 59 | ||
| BC120733.1 | - | 8 - 9 | 1474 | 40129656 | 59 | |
| BC137645.1 | - | 8 - 9 | 1474 | 8860664 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parp12
Genomic Map