| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600015473.mm.1 |
TC0600001970.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600015473.mm.1 |
NM_133925.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015473.mm.1 |
NM_133925.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015475.mm.1 |
TC0600001970.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600015475.mm.1 |
NM_133925.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015475.mm.1 |
NM_133925.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015471.mm.1 |
TC0600001970.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600015471.mm.1 |
NM_133925.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015471.mm.1 |
NM_133925.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015476.mm.1 |
TC0600001970.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600015476.mm.1 |
NM_133925.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015476.mm.1 |
NM_133925.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015474.mm.1 |
TC0600001970.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600015474.mm.1 |
NM_133925.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015474.mm.1 |
NM_133925.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015470.mm.1 |
TC0600001970.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600015470.mm.1 |
NM_133925.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015470.mm.1 |
NM_133925.2 |
Refseq |