Assay Details
| Gene Symbol: Irf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252382.1 | NP_001239311.1 | 6 - 7 | 955 | 71 | |
| NM_012057.4 | NP_036187.1 | 6 - 7 | 903 | 71 | ||
| GenBank mRNA | AF028725.1 | - | 6 - 7 | 795 | 71 | |
| AK150598.1 | - | 6 - 7 | 822 | 71 | ||
| AK151019.1 | - | 6 - 7 | 820 | 71 | ||
| AK152042.1 | - | 6 - 7 | 822 | 71 | ||
| AK156220.1 | - | 6 - 7 | 955 | 71 | ||
| AK171356.1 | - | 6 - 7 | 823 | 71 | ||
| EU401974.1 | - | 3 - 4 | 454 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Irf5
Genomic Map