Assay Details
| Gene Symbol: Ank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020332.4 | NP_065065.3 | 4 - 5 | 844 | 68 | |
| GenBank mRNA | AF001533.2 | - | 2 - 3 | 190 | 68 | |
| AF274752.1 | - | 4 - 5 | 547 | 68 | ||
| AK083135.1 | - | 4 - 5 | 845 | 68 | ||
| AK148070.1 | - | 4 - 5 | 617 | 68 | ||
| BC042644.1 | - | 4 - 5 | 769 | 68 | ||
| BC054379.1 | - | 4 - 5 | 751 | 6390764 | 68 | |
| DQ832285.1 | - | 4 - 5 | 844 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ank
Genomic Map