Assay Details
| Gene Symbol: Prps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026662.4 | NP_080938.1 | 2 - 3 | 417 | 68 | |
| GenBank mRNA | AK012819.1 | - | 2 - 3 | 449 | 68 | |
| AK019169.2 | - | 2 - 3 | 386 | 68 | ||
| AK029690.1 | - | 2 - 3 | 432 | 68 | ||
| AK045007.1 | - | 2 - 3 | 418 | 68 | ||
| BC024942.1 | - | 2 - 3 | 404 | 4022452 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prps2
Genomic Map