Assay Details
| Gene Symbol: Mmp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008611.4 | NP_032637.3 | 6 - 7 | 1016 | 129 | |
| GenBank mRNA | AK089234.1 | - | 6 - 7 | 987 | 129 | |
| AK089336.1 | - | 6 - 7 | 991 | 129 | ||
| AK089381.1 | - | 6 - 7 | 988 | 129 | ||
| AK089516.1 | - | 6 - 7 | 977 | 129 | ||
| AK137468.1 | - | 6 - 7 | 988 | 129 | ||
| AK154937.1 | - | 6 - 7 | 987 | 129 | ||
| AK155740.1 | - | 6 - 7 | 987 | 129 | ||
| AK155832.1 | - | 6 - 7 | 987 | 129 | ||
| AK155901.1 | - | 6 - 7 | 991 | 129 | ||
| AK170177.1 | - | 6 - 7 | 987 | 129 | ||
| AK170388.1 | - | 6 - 7 | 989 | 129 | ||
| AK170760.1 | - | 6 - 7 | 987 | 129 | ||
| AK171168.1 | - | 6 - 7 | 987 | 129 | ||
| AK171613.1 | - | 6 - 7 | 989 | 129 | ||
| BC042742.1 | - | 6 - 7 | 997 | 5032770 | 129 | |
| Y13342.1 | - | 6 - 7 | 914 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp8
Genomic Map